Learn More
Sino Biological SARS-CoV-2 Spike Antibody (Biotin), Rabbit PAb, Antigen Affinity Purified 20 µg
Catalog No. 5000749047
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant SARS-CoV-2 (2019-nCoV) Spike S1-His Recombiant Protein (HPLC-verified) (Catalog#40591-V08H; YP_009724390.1; Val16-Arg685) and then biotinylated. The specific IgG was purified by SARS-CoV-2 / 2019-nCoV Spike affinity chromatography and then biotinylated.
Catalog No. 50-007-49047 Supplier Sino Biological Supplier No. 40591T62B20
Produced in rabbits immunized with purified, recombinant SARS-CoV-2 (2019-nCoV) Spike S1-His Recombiant Protein (HPLC-verified) (Catalog#40591-V08H; YP_009724390.1; Val16-Arg685) and then biotinylated. The specific IgG was purified by SARS-CoV-2 / 2019-nCoV Spike affinity chromatography and then biotinylated.