Learn More
Sino Biological SARS-CoV-2 Spike Antibody (HRP), Mouse MAb 20 µg
Catalog No. 5000751261
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant SARS-CoV-2 Spike ( Catalog#40591-V05H1; YP_009724390.1; Val16-Arg685). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography and conjugated with horseradish-peroxidase (HRP).
Catalog No. 50-007-51261 Supplier Sino Biological Supplier No. 40591MM41H20
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant SARS-CoV-2 Spike ( Catalog#40591-V05H1; YP_009724390.1; Val16-Arg685). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography and conjugated with horseradish-peroxidase (HRP).