Learn More
Sino Biological SARS-CoV-2 Spike Antibody (HRP), Rabbit PAb, Antigen Affinity Purified 20 µg
Catalog No. 5000749898
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant SARS-CoV-2 (2019-nCoV) Spike S1-His Recombiant Protein (HPLC-verified) (Catalog#40591-V08H; YP_009724390.1; Val16-Arg685). The specific IgG was purified by SARS-CoV-2 / 2019-nCoV Spike affinity chromatography and conjugated with horseradish-peroxidase (HRP).
Catalog No. 50-007-49898 Supplier Sino Biological Supplier No. 40591T62H20
Produced in rabbits immunized with purified, recombinant SARS-CoV-2 (2019-nCoV) Spike S1-His Recombiant Protein (HPLC-verified) (Catalog#40591-V08H; YP_009724390.1; Val16-Arg685). The specific IgG was purified by SARS-CoV-2 / 2019-nCoV Spike affinity chromatography and conjugated with horseradish-peroxidase (HRP).