Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SARS-CoV-2 Spike Brazil (P.1) Ig Total Human ProcartaPlex™ Simplex Kit

Catalog No. 502576114
Encompass_Preferred
Change view
Click to view available options
Quantity:
96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-257-6114 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-257-6114 Supplier Invitrogen™ Supplier No. EPX01016014901
Only null left
Add to Cart
Add to Cart

The Human SARS-CoV-2 Spike Brazil (P.1) Ig Total Simplex ProcartaPlex Kit measures anti-SARS-CoV-2 Spike antibodies specific for the SARS-CoV-2 variant Lineage P.1 in serum and plasma.

The Human SARS-CoV-2 Spike Brazil (P.1) Ig Total Simplex ProcartaPlex Kit measures anti-SARS-CoV-2 Spike antibodies specific for the SARS-CoV-2 variant Lineage P.1 in serum and plasma. The detector antibody is an immunoglobulin (Ig) total antibody directly conjugated to phycoerythrin (PE) that cross-reacts with all Ig (IgG, IgM, and IgA) in serum and plasma. IgG is the most abundant of the immunoglobulins.

This kit is designed to be combinable with other Simplex kits from the Coronavirus Panel so that you can create your own multiplex panel that utilizes Luminex xMAP technology for protein detection/quantitation. When combining multiple Simplex kits (i.e., when you are not using a pre-configured Multiplex Panel), only one buffer kit (sold separately) is needed for each assay plate regardless of plex size.

ProcartaPlex immunoassays are based on the principles of a sandwich ELISA, using two highly specific antibodies binding to different epitopes of one protein to quantitate up to 80 protein targets simultaneously when using the FLEXMAP 3D or Luminex 200 instrument and up to 50 protein targets when using the MAGPIX instrument. ProcartaPlex assays require as little as 25 μL of plasma or serum, or 50 μL of cell culture supernatant, and just four hours to obtain analyzed results.

  • Flexible panels—design your own panels with Simplex kits to measure your own array of targets
  • More results per sample—measure up to 80 protein targets in a single 25–50 μL sample
  • Well-established Luminex technology—the most referenced multiplexing platform for protein detection and quantitation

The Luminex MagPlex superparamagnetic microsphere beads in the ProcartaPlex assay are internally dyed with precise proportions of red and infrared fluorophores to create 100 spectrally unique signatures that can be identified by the Luminex xMAP detec¬tion systems (Luminex 200, FLEXMAP 3D, and MAGPIX systems). Similar to a sandwich ELISA, the ProcartaPlex assay uses matched antibody pairs to identify the protein of interest. In a ProcartaPlex multiplex assay, each spectrally unique bead is labeled with antibodies specific for a single target protein, and bound proteins are identified with biotinylated antibodies and streptavidin–R-phycoerythrin (RPE). The conjugation of protein-specific antibodies to a distinct bead allows for analysis of multiple targets in a single well.

The most significant difference between a ProcartaPlex assay and ELISA is that the capture antibody in the ProcartaPlex assay is conjugated to a magnetic bead and not adsorbed to the microplate well, so the ProcartaPlex assay reagents are free-floating in the solution. For detection, the Luminex 200 instrument, for example, contains two lasers, one to distinguish the spectral signature of each bead and the second to quantify the amount of RPE fluorescence, which is proportional to the amount of protein present in the sample. ProcartaPlex multiplex assays can profile up to 80 times more target proteins using significantly less sample in the same time that it takes to perform a traditional sandwich ELISA.

ProcartaPlex Simplex kits provide the ability to create your own unique panel . More than 90% of ProcartaPlex Simplex targets can be combined, providing you with superior flexibility when creating your own multiplex panel.

ProcartaPlex Simplex kits are available across six species (human, mouse, rat, nonhuman primate, porcine, and canine).

TRUSTED_SUSTAINABILITY

Specifications

Product Type Simplex Kit
Quantity 96 Tests
Species Human
Combinability Combinable
Bead Region Spike Brazil (P0.1) [53 ]
Assay Sensitivity See Certificate of Analysis
Assay Range see product documentation
Format Simplex Kit
For Use With (Equipment) Luminex™ Instruments
Interassay CV Less than 15%
Intraassay CV Less than 15%
Protein Spike glycoprotein
Sample Type Serum, Plasma
Sample Volume Serum, Plasma: 25 μL (prediluted 1:1000)
Content And Storage 1 vial Capture Beads (50X)

Store at 2–8°C.
Shipping Condition Wet Ice
Research Area SARS-CoV-2 Ig Total
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.