Learn More
Genscript Corporation SARS-CoV-2 Spike E484 Neutra
Catalog No. 5000058369
Encompass
The product is specific for E484 site of wild-type RBD For specific mutant protein the product recognizes wild-type RBD RBD single mutant (N501Y) and RBD single mutant (K417N) but not recognizes RBD single mutant (E484K) and RBD triple mutant (K417N E484K and N501Y) as seen in Figure 2
Catalog No. 50-000-58369 Supplier Genscript Corporation Supplier No. A02109100
The product is specific for E484 site of wild-type RBD For specific mutant protein the product recognizes wild-type RBD RBD single mutant (N501Y) and RBD single mutant (K417N) but not recognizes RBD single mutant (E484K) and RBD triple mutant (K417N E484K and N501Y) as seen in Figure 2