Learn More
Sino Biological SARS-CoV-2 Omicron (BA.4/BA.5/BA.2.75) Spike RBD Specific Antibody, Chimeric MAb
Supplier:  Sino Biological 40589D00320
Encompass_Preferred
It is a chimeric monoclonal antibody combining the constant domains of the human IgG1 molecule with mouse variable regions. The variable region was obtained from a mouse immunized with purified, recombinant SARS-CoV-2 Omicron BA.4/BA.5 Spike ECD Protein. The antibody was produced using recombinant antibody technology.
Catalog No. 50-237-1141