Learn More
Sino Biological SARS-CoV-2 Spike MAb 20ug (Sino Biological)
Supplier:  Sino Biological 40589D00420
Encompass_Preferred
It is a chimeric monoclonal antibody combining the constant domains of the human IgG1 molecule with mouse variable regions. The variable region was obtained from a mouse immunized with purified, recombinant SARS-CoV-2 Alpha Spike ECD Protein. The antibody was produced using recombinant antibody technology.
Catalog No. 50-237-2664