Learn More
Sino Biological SARS-CoV-2 Spike MAb 20ug (Sino Biological)
Supplier:  Sino Biological 40591MM4520
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant SARS-CoV-2 Spike (Catalog#40591-V05H1; YP_009724390.1; Val16-Arg685). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-237-3852