Learn More
Sino Biological SARS-CoV-2 Spike MAb 20ul (Sino Biological)
Supplier:  Sino Biological 10837R00820
Encompass_Preferred
This antibody was obtained from a rabbit immunized with purified, recombinant SARS-CoV-2 Spike ( Catalog#40592-V05H; YP_009724390.1; Arg319-Phe541).
Catalog No. 50-237-4407