Learn More
Genscript Corporation SARS-CoV-2 Spike N501 Neutra
Catalog No. 5000057845
Encompass
The product is specific for N501 site of wild-type RBD For specific mutant protein the product recognizes wild-type RBD RBD single mutant (E484K) and RBD single mutant (K417N) but not recognizes RBD single mutant (N501Y) and RBD triple mutant (K417N E484K and N501Y) as seen in Figure 2
Catalog No. 50-000-57845 Supplier Genscript Corporation Supplier No. A021101
The product is specific for N501 site of wild-type RBD For specific mutant protein the product recognizes wild-type RBD RBD single mutant (E484K) and RBD single mutant (K417N) but not recognizes RBD single mutant (N501Y) and RBD triple mutant (K417N E484K and N501Y) as seen in Figure 2