Learn More
ACROBiosystems SARS-CoV-2 Spike NTD 25ug
Catalog No. 502534201
Encompass_Preferred
Biotinylated SARS-CoV-2 S1 protein NTD HisAvitag (S1D-C52E2) is expressed from human 293 cells (HEK293). It contains AA Val 16 - Phe 318 (Accession # QHD43416.1).
Catalog No. 50-253-4201 Supplier ACROBiosystems Supplier No. S1DC52E225UG
Biotinylated SARS-CoV-2 S1 protein NTD HisAvitag (S1D-C52E2) is expressed from human 293 cells (HEK293). It contains AA Val 16 - Phe 318 (Accession # QHD43416.1).