Learn More
ACROBiosystems ACROBiosystems SARS-CoV-2 Spike NTD (T19R, G142D, EF156-157del, R158G), His Tag (MALS verified)
Supplier:  ACROBiosystems S1DC52HH1MG
Encompass_Preferred
Recombinant Protein;1MG;SARS-CoV-2 Spike NTD, His Tag (S1D-C52Hh) is expressed from human 293 cells (HEK293). It contains AA Ser 13 - Leu 303 (Accession # QHD43416.1). The mutations (T19R, G142D, EF156-157del, R158G) were identified in the SARS-CoV-2 variants which emerged in India (known as B.1.617.2).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 34.6 kDa. The protein migrates as 50-60 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Anti-SARS-CoV-2 Spike NTD Antibody, Chimeric mAb (Cat. No. SPD-M121) at 1 μg/mL (100 μL/well) can bind SARS-CoV-2 Spike NTD, His Tag (Cat. No. S1D-C52Hh) with a linear range of 5-30 ng/mL (QC tested).
Catalog No. 50-210-6987