Learn More
Sino Biological SARS-CoV-2 Spike PAb 20ul (Sino Biological)
Supplier:  Sino Biological 40591T6220
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant SARS-CoV-2 (2019-nCoV) Spike S1-His Recombiant Protein (HPLC-verified) (Catalog#40591-V08H; YP_009724390.1; Val16-Arg685). The specific IgG was purified by SARS-CoV-2 / 2019-nCoV Spike affinity chromatography.
Catalog No. 50-237-2074