Learn More
Bon Opus Biosciences Recombinant SARS-CoV-2 Spike Protein (RBD) (C-His, HEK293)
SDP
Supplier:  Bon Opus Biosciences BP036100UG
Encompass_Preferred
Recombinant SARS-CoV-2 Spike Protein is produced by HEK293. The target gene encoding 319-532 is expressed with a 8His tag at the C terminus. Accession# P0DTC2. Bon Opus Cat# BP036
Catalog No. 50-203-1808