Learn More
Genscript Corporation SARS-CoV-2 Spike protein (RBD, E484K, K417T, N501Y, Avi & His Tag)
Supplier:  Genscript Corporation Z03600100
Encompass
SARS-CoV-2 (Severe Acute Respiratory Syndrome Coronavirus 2) also known as 2019-nCoV (2019 Novel Coronavirus) is a virus that causes illnesses ranging from the common cold to severe diseases. Lineage P.1, also known as the Brazil(ian) variant is one of the variants of SARS-CoV-2. This variant has 17 amino acid changes, ten of which are in its spike protein, including these three designated to be of particular concern: N501Y, E484K and K417T. SARS-CoV-2 Spike protein (RBD, E484K, K417T, N501Y, Avi & His Tag) carries a Avi Tag followed by a polyhistidine tag at the C-terminus. It has high binding affinity to human chimeric ACE2.
Catalog No. NC1960555