Learn More
ACROBiosystems Biotinylated SARS-CoV-2 Spike Trimer, His,Avitag™ (BA.2/Omicron) (SPN-C82Er) is expressed from human 293 cells (HEK293). It contains AA Val 16 - Pro 1213 (Accession # QHD43416.1 (T19I, LPP24-26del, A27S, G142D, V213G, G339D, S371F, S373P, S
Catalog No. 502253557
Encompass_Preferred
It's been reported that SARS-CoV-2 can infect the human respiratory epithelial cells through interaction with the human ACE2 receptor. The spike protein is a large type I transmembrane protein containing two subunits, S1 and S2. S1 mainly contains a receptor binding domain (RBD), which is responsible for recognizing the cell surface receptor. S2 contains basic elements needed for the membrane fusion.The S protein plays key parts in the induction of neutralizing-antibody and T-cell responses, as well as protective immunity.
This item is not returnable. View return policy
Catalog No. 50-225-3557 Supplier ACROBiosystems Supplier No. SPNC82ER25UG
 Restricted Item
Add to Cart
Add to Cart
It's been reported that SARS-CoV-2 can infect the human respiratory epithelial cells through interaction with the human ACE2 receptor. The spike protein is a large type I transmembrane protein containing two subunits, S1 and S2. S1 mainly contains a receptor binding domain (RBD), which is responsible for recognizing the cell surface receptor. S2 contains basic elements needed for the membrane fusion.The S protein plays key parts in the induction of neutralizing-antibody and T-cell responses, as well as protective immunity.