Learn More
Sino Biological SARS-CoV-2 Spike S1+S2 (E484K, F565L, D614G, V1176F) Protein (ECD, His Tag)
Supplier:  Sino Biological 40589V08B19100
Encompass_Preferred
A DNA sequence encoding the SARS-CoV-2 Spike S1+S2 (YP_009724390.1, with mutations E484K, F565L, D614G, V1176F) (Met1-Pro1213) was expressed with a polyhistidine tag at the C-terminus. The mutations were identified in the SARS-CoV-2 variant (known as variant P.2) which emerged in the Brazil.
Catalog No. 50-237-3421