Learn More
Sino Biological SARS-CoV-2 Spike Protein 100ug (Sino Biological)
Supplier:  Sino Biological 40592V08H138100
Encompass_Preferred
A DNA sequence encoding the SARS-CoV-2 (BA.3) Spike RBD (YP_009724390.1, with mutation G339D, S371F, S373P, S375F, D405N, K417N, N440K, G446S, S477N, T478K, E484A, Q493R, Q498R, N501Y, Y505H) (Arg319-Lys537) was expressed with a polyhistidine tag at the C-terminus. The mutations were identified in the SARS-CoV-2 variant (known as variant BA.3).
Catalog No. 50-237-3755