Learn More
Sino Biological SARS-CoV-2 Spike Protein 100ug (Sino Biological)
Supplier:  Sino Biological 10369MM1520
Encompass_Preferred
A DNA sequence encoding the SARS-CoV-2 Spike RBD (YP_009724390.1, with mutations Y449H, E484K, N501Y) (Arg319-Phe541) was expressed with a polyhistidine tag at the C-terminus. The mutations were identified in the SARS-CoV-2 variant (known as variant C.1.2) which emerged in the South Africa.
Catalog No. 50-237-4220