Learn More
Genscript Corporation SARS-CoV-2 Spike protein RBD
Catalog No. 5000059129
Encompass
SARS-CoV-2 (Severe Acute Respiratory Syndrome Coronavirus 2) also known as 2019-nCoV (2019 Novel Coronavirus) is a virus that causes illnesses ranging from the common cold to severe diseases SARS-CoV-2 Spike Protein is composed of S1 domain and S2 domain S1 contains a receptor-binding domain (RBD) that can specifically bind to angiotensin-converting enzyme 2 (ACE2) the receptor on target cells It is believed that SARS-CoV-2 Spike Protein (RBD) has potential value for the diagnosis of the virus
Catalog No. 50-000-59129 Supplier Genscript Corporation Supplier No. Z03483100
SARS-CoV-2 (Severe Acute Respiratory Syndrome Coronavirus 2) also known as 2019-nCoV (2019 Novel Coronavirus) is a virus that causes illnesses ranging from the common cold to severe diseases SARS-CoV-2 Spike Protein is composed of S1 domain and S2 domain S1 contains a receptor-binding domain (RBD) that can specifically bind to angiotensin-converting enzyme 2 (ACE2) the receptor on target cells It is believed that SARS-CoV-2 Spike Protein (RBD) has potential value for the diagnosis of the virus