Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SARS-CoV-2 Spike Protein (RBD) (aa330-524), His Tag Recombinant Protein

Catalog No. PIRP87674
Change view
Click to view available options
Quantity:
1 mg
100 μg
200 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIRP87674 100 μg
PIRP87683 200 μg
PIRP87684 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. PIRP87674 Supplier Invitrogen™ Supplier No. RP87674
Only null left
Add to Cart
Add to Cart

Recombinant Protein

RP-87674 has been tested in indirect ELISA. Reconstitution: It is recommend that the vial be centrifuged briefly (∽30 seconds) before opening the tube in order to bring the contents to the bottom. Then, reconstitute the protein with 400 μL of deionized sterile water to obtain a final concentration of 0.5 mg/mL. Let the tube sit on ice for ∽30 minutes with gentle pipetting periodically.

SARS-CoV-2 (Severe Acute Respiratory Syndrome Coronavirus 2) also known as 2019-nCoV (2019 Novel Coronavirus) is a virus that causes illnesses ranging from the common cold symptoms to severe effects including death and respiratory distress. The CoV Spike (S) protein plays the most important roles in viral attachment, fusion and entry, and serves as a target for development of antibodies, entry inhibitors and vaccines. Of note is the Spike protein receptor-binding domain (RBD, S-RBD) in SARS-CoV-2 Spike protein which binds strongly to human and bat angiotensin-converting enzyme 2 (ACE2) receptors. SARS-CoV-2 RBD exhibits significantly higher binding affinity to ACE2 receptor than SARS-CoV RBD in comparison. S1 contains a receptor-binding domain (RBD) that can specifically bind to angiotensin-converting enzyme 2 (ACE2), the receptor on target cells.
TRUSTED_SUSTAINABILITY

Specifications

Concentration Conc. Not Determined
For Use With (Application) Control, ELISA Standard, Binding Assay
Formulation PBS with no preservative
Molecular Weight (g/mol) 27 kDa
Name SARS-CoV-2 Spike Protein (RBD) (aa330-524), His Tag
Purification Method Purified
Quantity 100 μg
Source E. coli
Storage Requirements -20°C or -80°C if preferred
Regulatory Status RUO
Gene Alias Coronavirus; COVID; Covid 19; COVID19; COVID-19; SARS; SARS Coronavirus; SARS CoV; SARS CoV2; SARS CoV-2; SARS Spike Protein; SARS-CoV2; SARS-CoV-2
Common Name SARS-CoV-2 Spike Protein RBD
Product Type Protein
Conjugate Unconjugated
Recombinant Recombinant
Protein Tag His-tag
Sequence SARS-CoV-2 Spike Protein (s-RBD) contains amino acids 330-524 of the SARS-CoV-2 Spike protein with a His-tag
Content And Storage -20°Cor to +80°C if preferred
Expression System E. coli
Form Lyophilized
Purity or Quality Grade >95% by SDS-PAGE under reducing conditions
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.