Learn More
Invitrogen™ SARS-CoV-2 Spike Protein S1 Fc Chimera Recombinant Protein
Description
It is recommended that the protein be aliquoted for optimal storage. It is recommended that sterile water (400 μL) be added to the vial to prepare a stock solution of 0.25 mg/mL. Concentration is measured by UV.
Specifications
Specifications
| For Use With (Application) | Control, ELISA Standard, Binding Assay |
| Format | Lyophilized |
| Formulation | PBS with 0.01% mannitol, 5-8% trehalose, 0.01% Tween 80 and no preservative; pH 7.4 |
| Molecular Weight (g/mol) | 101.7 kDa |
| Name | SARS-CoV-2 Spike Protein S1 Fc Chimera |
| pH Range | 7.4 |
| Purification Method | Purified |
| Quantity | 100 μg |
| Storage Requirements | -20°C, Avoid Freeze/Thaw Cycles |
| Regulatory Status | RUO |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.