Learn More
Genscript Corporation SARS-CoV-2 Spike protein S1 O
Catalog No. 5000056661
Encompass
SARS-CoV-2 (Severe Acute Respiratory Syndrome Coronavirus 2) also known as 2019-nCoV (2019 Novel Coronavirus) is a virus that causes illnesses ranging from the common cold to severe diseases Recently the new B 1 1 529 variant was confirmed in South Africa and preliminary evidence suggests an increased risk of reinfection with this variant The B 1 1 529 variant was first reported to WHO on 24 November 2021 and WHO has designated this variant as a VOC (Variant of Concern) named Omicron There are more than 30 mutations in the spike protein
Catalog No. 50-000-56661 Supplier Genscript Corporation Supplier No. Z037291
SARS-CoV-2 (Severe Acute Respiratory Syndrome Coronavirus 2) also known as 2019-nCoV (2019 Novel Coronavirus) is a virus that causes illnesses ranging from the common cold to severe diseases Recently the new B 1 1 529 variant was confirmed in South Africa and preliminary evidence suggests an increased risk of reinfection with this variant The B 1 1 529 variant was first reported to WHO on 24 November 2021 and WHO has designated this variant as a VOC (Variant of Concern) named Omicron There are more than 30 mutations in the spike protein