Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SARS-CoV-2 Spike Protein S2 Fc Chimera Recombinant Protein

Catalog No. PIRP88439
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIRP88439 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIRP88439 Supplier Invitrogen™ Supplier No. RP88439
Add to Cart
Add to Cart

Recombinant Protein

It is recommended that the protein be aliquoted for optimal storage. It is recommended that sterile water (400 μL) be added to the vial to prepare a stock solution of 0.25 mg/mL. Concentration is measured by UV.

SARS-CoV-2 (Severe Acute Respiratory Syndrome Coronavirus 2) also known as 2019-nCoV (2019 Novel Coronavirus) is a virus that causes illnesses ranging from the common cold symptoms to severe effects including death and respiratory distress. SARS-CoV-2 Spike Protein is composed of S1 domain and S2 domain (Spike Protein). The S2 domain plays a role in fusion. Further, S proteins plays an important role in the induction of neutralizing-antibodies and T-cell responses, as well as protective immunity. CoV diversity is reflected in the variable Spike proteins (S proteins), which have evolved into forms differing in their receptor interactions and their response to various environmental triggers of virus-cell membrane fusion.
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Control, Neutralization
Format Lyophilized
Formulation PBS with 0.01% mannitol, 5-8% trehalose, 0.01% Tween 80 and no preservative; pH 7.4
Molecular Weight (g/mol) 84.66 kDa
Name SARS-CoV-2 Spike Protein S2 Fc Chimera
pH Range 7.4
Purification Method Purified
Quantity 100 μg
Storage Requirements -20°C, Avoid Freeze/Thaw Cycles
Regulatory Status RUO
Endotoxin Concentration <1.0 EU/μg
Gene Alias Coronavirus; COVID; Covid 19; COVID19; COVID-19; COVID-19 S2; COVID19 Spike S2; SARS; SARS Coronavirus; SARS CoV; SARS CoV2; SARS CoV-2; SARS Spike Protein; SARS-CoV2; SARS-CoV-2
Common Name SARS-CoV-2 Spike Protein S2
Product Type Protein
Conjugate Unconjugated
Recombinant Recombinant
Protein Tag Fc-tag
Sequence SARS-CoV-2 spike S2, amino acids Ser686-Pro1213 (Accession # YP_009724390.1) with a C-terminal human IgG1 Fc region
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Expression System HEK293 cells
Form Lyophilized
Purity or Quality Grade >95% as determined by SDS-PAGE.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.