Learn More
Sino Biological SARS-CoV-2 (2019-nCoV) Spike Antibody, Rabbit PAb, Antigen Affinity Purified
Supplier:  Sino Biological 40589T62100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant SARS-CoV-2 / 2019-nCoV Spike Protein (S1+S2 ECD)(Catalog#40589-V08B1; YP_009724390.1; Val16-Pro1213). The specific IgG was purified by SARS-CoV-2 / 2019-nCoV Spike affinity chromatography.
Catalog No. NC1821294