Learn More
ACROBiosystems SARS-CoV-2 Spike RBD Protein, His Tag (XBB.1.5 Omicron), 100 μg
Supplier: ACROBiosystems SPDC5242100UG
Recombinant SARS-CoV-2 spike receptor-binding domain (RBD) from the XBB.1.5 (Omicron) variant, His-tagged and MALS-verified. Supplied lyophilized for assay development, antibody characterization, and binding studies.
- Expression system: Human 293 cells (HEK293).
- Purity: >95% by SDS-PAGE; >90% by SEC-MALS.
- Molecular weight: calculated 26.6 kDa; migrates 35-42 kDa on SDS-PAGE due to glycosylation.
- Format: lyophilized from PBS (pH 7.4) with trehalose.
- Storage: stable 12 months at -20°C to -70°C (lyophilized); 3 months at -70°C after reconstitution.
- Tag: C-terminal polyhistidine tag.
- QC: verified by SEC-MALS.
- Size: 100 μg.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.