Learn More
ACROBiosystems SARS-CoV-2 Spike RBD Protein, His Tag (XBB.1.5/Omicron), 1 mg (2 x 500 μg)
Catalog No. 502357295
Encompass_Preferred
SARS-CoV-2 Spike RBD Protein (XBB.1.5/Omicron) is a lyophilized recombinant receptor-binding domain fragment expressed in human HEK293 cells and supplied as a 1 mg pack (2 x 500 μg). The polyhistidine-tagged protein is MALS-verified, low endotoxin, and intended for use as an ELISA antigen, assay standard, or control in binding and serological assays.
  • Expressed in human HEK293 cells
  • Polyhistidine tag at the C-terminus for purification
  • High purity: >95% by SDS-PAGE; >90% by SEC-MALS
  • Calculated molecular weight 26.6 kDa; migrates 35-42 kDa on SDS-PAGE due to glycosylation
  • Lyophilized from PBS, pH 7.4, with trehalose
  • Low endotoxin: <1.0 EU per μg
  • MALS verified for molecular mass confirmation
  • Supplied as two 500 μg vials (total 1 mg)
  • Stable lyophilized at -20°C to -70°C for up to 12 months; stable for 3 months at -70°C after reconstitution
Catalog No. 50-235-7295 Supplier ACROBiosystems Supplier No. SPDC52421MG
SARS-CoV-2 Spike RBD Protein (XBB.1.5/Omicron) is a lyophilized recombinant receptor-binding domain fragment expressed in human HEK293 cells and supplied as a 1 mg pack (2 x 500 μg). The polyhistidine-tagged protein is MALS-verified, low endotoxin, and intended for use as an ELISA antigen, assay standard, or control in binding and serological assays.
  • Expressed in human HEK293 cells
  • Polyhistidine tag at the C-terminus for purification
  • High purity: >95% by SDS-PAGE; >90% by SEC-MALS
  • Calculated molecular weight 26.6 kDa; migrates 35-42 kDa on SDS-PAGE due to glycosylation
  • Lyophilized from PBS, pH 7.4, with trehalose
  • Low endotoxin: <1.0 EU per μg
  • MALS verified for molecular mass confirmation
  • Supplied as two 500 μg vials (total 1 mg)
  • Stable lyophilized at -20°C to -70°C for up to 12 months; stable for 3 months at -70°C after reconstitution