Learn More
ACROBiosystems Biotinylated SARS-CoV-2 Spike RBD (XBB.1.5 Omicron), His-Avitag, 25 μg
Supplier: ACROBiosystems SPDC82Q325UG
Biotinylated SARS-CoV-2 Spike receptor-binding domain (RBD) from the XBB.1.5 (Omicron) lineage, supplied as a lyophilized 25 μg vial. The construct comprises residues Arg 319-Lys 537, includes a C-terminal His tag and an Avi tag that is enzymatically biotinylated, and is expressed in HEK293 cells. Designed for ELISA, binding assays, and reagent development.
- Lyophilized 25 μg vial for assay use.
- Contains residues Arg 319-Lys 537 (RBD).
- Expressed in HEK293 (human) cells.
- C-terminal His tag and Avi tag; site-specific enzymatic biotinylation.
- Purity: >95% by SDS-PAGE, >90% by SEC-MALS.
- Calculated molecular weight 28.3 kDa; migrates ~35-41 kDa by SDS-PAGE due to glycosylation.
- Formulated in PBS, pH 7.4, with trehalose; 0.22 μm filtered prior to lyophilization.
- Store lyophilized at -20°C or lower.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.