Learn More
ACROBiosystems SARS-CoV-2 Spike RBD 25ug
Catalog No. 502534148
Encompass_Preferred
Biotinylated SARS-CoV-2 Spike RBD Protein HisAvitag (BA.2.86/EPI_ISL_18114953) (SPD-C82Q6) is expressed from human 293 cells (HEK293). It contains AA Arg 319 - Lys 537 (Accession # QHD43416.1 (I332VG339HK356TS371FS373PS375FT376AR403KD405NR408SK417NN440KV445HG446SN450DL452WN460KS477NT478KN481KV483delE484KF486PQ498RN501YY505H)). The spike mutations are identified on the SARS-CoV-2 Omicron variant (Pango lineage: BA.2.86/Denmark/EPI_ISL_18114953).
Catalog No. 50-253-4148 Supplier ACROBiosystems Supplier No. SPDC82Q625UG
Biotinylated SARS-CoV-2 Spike RBD Protein HisAvitag (BA.2.86/EPI_ISL_18114953) (SPD-C82Q6) is expressed from human 293 cells (HEK293). It contains AA Arg 319 - Lys 537 (Accession # QHD43416.1 (I332VG339HK356TS371FS373PS375FT376AR403KD405NR408SK417NN440KV445HG446SN450DL452WN460KS477NT478KN481KV483delE484KF486PQ498RN501YY505H)). The spike mutations are identified on the SARS-CoV-2 Omicron variant (Pango lineage: BA.2.86/Denmark/EPI_ISL_18114953).