Learn More
Rockland Immunochemicals SARS-CoV-2 Spike (RBD) 25µL
SDP
Supplier:  Rockland Immunochemicals 600401MS8S
Encompass
Anti-SARS-CoV-2 Spike protein purified antibody has been tested for use in ELISA and shows reactivity to recombinant SARS COV-2, S1 subunit protein and recombinant SARS COV-2 whole spike protein.  Specific conditions for reactivity should be optimized by the end user.
Catalog No. 50-202-4102