Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ACROBiosystems SARS-CoV-2 Spike RBD Protein (XBB/Omicron), C-terminal His tag, HEK293, lyophilized, 1 mg

Catalog No. NC2219091
Encompass_Preferred

Recombinant SARS-CoV-2 spike receptor-binding domain (RBD) representing the XBB/Omicron lineage, expressed in human HEK293 cells with a C-terminal polyhistidine tag. MALS-verified and supplied lyophilized for use in ELISA and antibody/ACE2 binding assays.

  • Expression system: HEK293 (human 293) cells.
  • Sequence: Arg 319-Lys 537 (RBD).
  • Variant: XBB/Omicron mutations present.
  • Tag: C-terminal polyhistidine (His) tag.
  • Molecular weight: ~30-40 kDa (SEC-MALS verified).
  • Purity: >95% by SDS-PAGE; >90% by SEC-MALS.
  • Formulation: lyophilized from 0.22 μm filtered PBS, pH 7.4, with trehalose.
  • Storage: long-term lyophilized at -20°C or below; reconstituted storage at -70°C (sterile) for short term.
  • Applications: bioactivity ELISA and antibody/ACE2 binding assays.

This item is not returnable. View return policy
Catalog No. NC2219091 Supplier ACROBiosystems Supplier No. SPDC52411MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant SARS-CoV-2 spike receptor-binding domain (RBD) representing the XBB/Omicron lineage, expressed in human HEK293 cells with a C-terminal polyhistidine tag. MALS-verified and supplied lyophilized for use in ELISA and antibody/ACE2 binding assays.

  • Expression system: HEK293 (human 293) cells.
  • Sequence: Arg 319-Lys 537 (RBD).
  • Variant: XBB/Omicron mutations present.
  • Tag: C-terminal polyhistidine (His) tag.
  • Molecular weight: ~30-40 kDa (SEC-MALS verified).
  • Purity: >95% by SDS-PAGE; >90% by SEC-MALS.
  • Formulation: lyophilized from 0.22 μm filtered PBS, pH 7.4, with trehalose.
  • Storage: long-term lyophilized at -20°C or below; reconstituted storage at -70°C (sterile) for short term.
  • Applications: bioactivity ELISA and antibody/ACE2 binding assays.

This item is not returnable. View return policy
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.