Learn More
Sino Biological SARS-CoV-2 (2019-nCoV) Spike RBD-AVI & His Recombinant Protein, Biotinylated (HPLC-verified)
Supplier:  Sino Biological 40592V27HB20
Encompass_Preferred
A DNA sequence encoding the SARS-CoV-2 (2019-nCoV) Spike RBD Protein (YP_009724390.1) (Arg319-Phe541) was expressed with a c-terminal AVI tagged polyhistidine tag at the C-terminus. The purified protein was biotinylated in vitro.
Catalog No. 50-211-5534