Learn More
ACROBiosystems SARS-CoV-2 Spike RBD Protein, His Tag (BQ.1.1/Omicron) - 1 mg
Supplier: ACROBiosystems SPDC52401MG
Recombinant SARS-CoV-2 spike receptor binding domain (RBD) expressed in human HEK293 cells with a C-terminal His tag. Suitable for ACE2-binding assays, antibody characterization, and serology research.
- Expressed in human HEK293 cells
- C-terminal polyhistidine (His) tag
- Available as 1 mg (500 μg x 2)
- Purity: >95% by SDS-PAGE; >90% by SEC-MALS
- Calculated molecular weight 26.6 kDa; migrates 33-37 kDa on SDS-PAGE
- Contains RBD residues Arg319-Lys537 with BQ.1.1 Omicron mutations
- Intended for research use in ACE2 interaction and antibody studies
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.