Learn More
Sino Biological SARS-CoV-2 (2019-nCoV) Spike RBD (K417T, E484K, N501Y) Protein (His Tag) 100ug
Supplier:  Sino Biological 40592V08H86100
Encompass_Preferred
A DNA sequence encoding the SARS-CoV-2 (2019-nCoV) Spike RBD (YP_009724390.1, with mutations K417T, E484K, N501Y) (Arg319-Phe541) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-211-6502