Learn More
Sino Biological SARS-CoV-2 Spike RBD (non-RBM) Peptide Pool 1 vial (50 μg/peptide)
Catalog No. 5000696231
Encompass_Preferred
The SARS-CoV-2 Spike RBD (non-RBM) Peptide Pool, consisting mainly of 15-mer sequences with 11 amino acids (aa) overlap.
Catalog No. 50-006-96231 Supplier Sino Biological Supplier No. PP002B1
The SARS-CoV-2 Spike RBD (non-RBM) Peptide Pool, consisting mainly of 15-mer sequences with 11 amino acids (aa) overlap.