Learn More
Sino Biological SARS-CoV-2 (2019-nCoV) Spike RBD(K417N)-His Recombinant Protein 100ug
Supplier:  Sino Biological 40592V08H59100
Encompass_Preferred
A DNA sequence encoding the SARS-CoV-2 (2019-nCoV) Spike RBD(K417N)-His Recombinant Protein (YP_009724390.1) (Arg319-Phe541(K417N)) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-211-5922