Learn More
Sino Biological SARS-CoV-2 (2019-nCoV) Spike RBD(S477N)-His Recombinant Protein
Supplier:  Sino Biological 40592V08H46100
Encompass_Preferred
A DNA sequence encoding the SARS-CoV-2 (2019-nCoV) Spike RBD(S477N)-His Recombinant Protein (YP_009724390.1) (Arg319-Phe541(S477N)) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-211-5911