Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC SARS-CoV-2 spike receptor-binding domain (Omicron, B.1.1.529) | >95.0% | 10 UG
SDP

Catalog No. 5000293772
Encompass_Preferred

Recombinant SARS-CoV-2 spike receptor-binding domain (Omicron B.1.1.529), expressed in mammalian cells and purified for use in biochemical and immunological assays. The protein carries a C-terminal His tag, is supplied as a sterile, filtered PBS solution, and is intended for applications such as ELISA, binding studies, and antibody characterization.

  • Expressed in HEK293 cells with a C-terminal His tag.
  • Represents the Omicron (B.1.1.529) variant of the spike RBD.
  • Purity greater than 95% by reducing SDS-PAGE.
  • Supplied as a 0.22 μm filtered PBS solution, pH 7.4.
  • Store at -80°C; shipped on dry ice; avoid repeated freeze-thaw cycles.
  • Suitable for ELISA, binding assays, and antibody characterization.

Catalog No. 50-002-93772 Supplier Medchemexpress LLC Supplier No. HYP7444710UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant SARS-CoV-2 spike receptor-binding domain (Omicron B.1.1.529), expressed in mammalian cells and purified for use in biochemical and immunological assays. The protein carries a C-terminal His tag, is supplied as a sterile, filtered PBS solution, and is intended for applications such as ELISA, binding studies, and antibody characterization.

  • Expressed in HEK293 cells with a C-terminal His tag.
  • Represents the Omicron (B.1.1.529) variant of the spike RBD.
  • Purity greater than 95% by reducing SDS-PAGE.
  • Supplied as a 0.22 μm filtered PBS solution, pH 7.4.
  • Store at -80°C; shipped on dry ice; avoid repeated freeze-thaw cycles.
  • Suitable for ELISA, binding assays, and antibody characterization.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.