Learn More
ACROBiosystems SARS-CoV-2 Spike S1 100ug
Catalog No. 502534199
Encompass_Preferred
SARS-CoV-2 S1 protein (N501Y) His Tag (S1N-C52Hg) is expressed from human 293 cells (HEK293). It contains AA Val 16 - Arg 685 (Accession # QHD43416.1 (N501Y)). N501Y mutaion was identified in multiple SARS-CoV-2 Variants of Concerns (VOCs) i.e. the Alpha variant (Pango lineage: B.1.1.7; other names: 20I/501Y.V1 or VOC 202012/01) the Beta variant (Pango lineage: B.1.351; other names: 20H/501Y.V2) the Gamma variant (Pango lineage: P.1; other names: 20J/501Y.V3).
Catalog No. 50-253-4199 Supplier ACROBiosystems Supplier No. S1NC52HG100UG
SARS-CoV-2 S1 protein (N501Y) His Tag (S1N-C52Hg) is expressed from human 293 cells (HEK293). It contains AA Val 16 - Arg 685 (Accession # QHD43416.1 (N501Y)). N501Y mutaion was identified in multiple SARS-CoV-2 Variants of Concerns (VOCs) i.e. the Alpha variant (Pango lineage: B.1.1.7; other names: 20I/501Y.V1 or VOC 202012/01) the Beta variant (Pango lineage: B.1.351; other names: 20H/501Y.V2) the Gamma variant (Pango lineage: P.1; other names: 20J/501Y.V3).