Learn More
ACROBiosystems SARS-CoV-2 Spike RBD-coupled Magnetic Beads
Catalog No. 502331483
Encompass_Preferred
SARS-CoV-2 Spike RBD protein is expressed from human 293 cells (HEK293). It contains AA Arg 319 - Phe 541(Accession # QHD43416.1).
Catalog No. 50-233-1483 Supplier ACROBiosystems Supplier No. MBSK0022MG
SARS-CoV-2 Spike RBD protein is expressed from human 293 cells (HEK293). It contains AA Arg 319 - Phe 541(Accession # QHD43416.1).