Learn More
Sino Biological SARS-CoV-2 (2019-nCoV) Spike S1-His Recombinant Protein, Biotinylated
Supplier:  Sino Biological 40591V08HB20
Encompass_Preferred
A DNA sequence encoding the SARS-CoV-2 (2019-nCoV) spike protein S1 Subunit (YP_009724390.1) (Val16-Arg685) was expressed with a polyhistidine tag at the C-terminus. The purified protein was biotinylated in vitro.
Catalog No. 50-211-5511
 Restricted Item