Learn More
Sino Biological SARS-CoV-2 (2019-nCoV) Spike S1 NTD-His & AVI Recombinant Protein, Biotinylated
Supplier:  Sino Biological 40591V49HB20
Encompass_Preferred
A DNA sequence encoding the SARS-CoV-2 (2019-nCoV) Spike S1 NTD (YP_009724390.1) (Met1-Ser305) was expressed with a C-terminal polyhistidine tag followed by an AVI tag. The purified protein was biotinylated in vitro.
Catalog No. 50-211-5615