Learn More
Sino Biological SARS-CoV-2 Spike S1 Peptide Pool 1 vial (50 μg/peptide)
Catalog No. 5000696169
Encompass_Preferred
The SARS-CoV-2 Spike S1 Peptide Pool, consisting mainly of 15-mer sequences with 11 amino acids (aa) overlap.
Catalog No. 50-006-96169 Supplier Sino Biological Supplier No. PP003A1
The SARS-CoV-2 Spike S1 Peptide Pool, consisting mainly of 15-mer sequences with 11 amino acids (aa) overlap.