Learn More
Sino Biological SARS-CoV-2 (2019-nCoV) Spike S1(A222V, D614G)-His Recombinant Protein 100ug
Supplier:  Sino Biological 40591V08H4100
Encompass_Preferred
A DNA sequence encoding the SARS-CoV-2 (2019-nCoV) Spike S1(A222V, D614G)-His Recombinant Protein (YP_009724390.1) (Met1-Arg685(A222V, D614G)) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-211-6457
 Restricted Item