Learn More
Abcam SARS-CoV-2 Spike S1+S2 (del H69+V70+Y144 mut N501Y+A570D+D614G+P681H+T716I+S982A+D1118H) protein, 100UG
Catalog No. 502679261
Encompass_Preferred
SARS-CoV-2 Spike S1+S2 (del H69+V70+Y144 mut N501Y+A570D+D614G+P681H+T716I+S982A+D1118H) protein is a SARS-CoV-2 Fragment protein, in the 1 to 1213 aa range, expressed in Baculovirus infected insect, with >85% purity, < 1 EU/µg endotoxin level and suitable for SDS-PAGE, FuncS.
Protein/ Peptide/ Enzyme Synonyms: 2, S, Spike glycoprotein, S glycoprotein, E2, Peplomer protein
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-267-9261 Supplier Abcam Supplier No. AB289619100UG
SARS-CoV-2 Spike S1+S2 (del H69+V70+Y144 mut N501Y+A570D+D614G+P681H+T716I+S982A+D1118H) protein is a SARS-CoV-2 Fragment protein, in the 1 to 1213 aa range, expressed in Baculovirus infected insect, with >85% purity, < 1 EU/µg endotoxin level and suitable for SDS-PAGE, FuncS.
Protein/ Peptide/ Enzyme Synonyms: 2, S, Spike glycoprotein, S glycoprotein, E2, Peplomer protein
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement