Learn More
Abcam SARS-CoV-2 Spike S1+S2 (mut L18F+T20N+P26S+D138Y+R190S+K417T+E484K+N501Y+D614G+H655Y+T1027I), 100UG
Catalog No. 502679258
Encompass_Preferred
SARS-CoV-2 Spike S1+S2 (mut L18F+T20N+P26S+D138Y+R190S+K417T+E484K+N501Y+D614G+H655Y+T1027I) is a SARS-CoV-2 Fragment protein, in the 1 to 1213 aa range, expressed in Baculovirus infected insect, with >85% purity, < 1 EU/µg endotoxin level and suitable for SDS-PAGE, FuncS.
Protein/ Peptide/ Enzyme Synonyms: 2, S, Spike glycoprotein, S glycoprotein, E2, Peplomer protein
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-267-9258 Supplier Abcam Supplier No. AB289622100UG
SARS-CoV-2 Spike S1+S2 (mut L18F+T20N+P26S+D138Y+R190S+K417T+E484K+N501Y+D614G+H655Y+T1027I) is a SARS-CoV-2 Fragment protein, in the 1 to 1213 aa range, expressed in Baculovirus infected insect, with >85% purity, < 1 EU/µg endotoxin level and suitable for SDS-PAGE, FuncS.
Protein/ Peptide/ Enzyme Synonyms: 2, S, Spike glycoprotein, S glycoprotein, E2, Peplomer protein
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement