Learn More
Abcam SARS-CoV-2 Spike S1+S2 (mutated D80A + K417N + E484K + N501Y + D614G + A701V) protein (Active), 100UG
Catalog No. 502679260
Encompass_Preferred
SARS-CoV-2 Spike S1+S2 (mutated D80A + K417N + E484K + N501Y + D614G + A701V) protein (Active) is a SARS-CoV-2 Fragment protein, in the 1 to 1213 aa range, expressed in Baculovirus infected insect, with =90% purity, < 1 EU/µg endotoxin level and suitable for SDS-PAGE, FuncS.
Protein/ Peptide/ Enzyme Synonyms: 2, S, Spike glycoprotein, S glycoprotein, E2, Peplomer protein
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-267-9260 Supplier Abcam Supplier No. AB289620100UG
SARS-CoV-2 Spike S1+S2 (mutated D80A + K417N + E484K + N501Y + D614G + A701V) protein (Active) is a SARS-CoV-2 Fragment protein, in the 1 to 1213 aa range, expressed in Baculovirus infected insect, with =90% purity, < 1 EU/µg endotoxin level and suitable for SDS-PAGE, FuncS.
Protein/ Peptide/ Enzyme Synonyms: 2, S, Spike glycoprotein, S glycoprotein, E2, Peplomer protein
Conjugation: Unconjugated
The product is subject to the following: Abcam Restricted Use Statement