Learn More
ACROBiosystems ACROBiosystems SARS-CoV-2 Spike Trimer (P.1) Coupled Magnetic Beads
Catalog No. 502107050
Encompass_Preferred
Beads;10MG;The magnetic beads technology makes use of the easy and efficient collection of beads in magnetic field to facilitate antibody purification in a simple workflow of “bind-wash-elute”. In contrast to common separation techniques, this method does not require columns or centrifugation, and is therefore ideal in high-throughput applications. ;Immobilized 40μg S protein trimer/1mg beads can bind the Anti-S1 Antibody with an EC50 of 0.5720μg/mL (QC tested).
Catalog No. 50-210-7050 Supplier ACROBiosystems Supplier No. MBSK03110MG
Beads;10MG;The magnetic beads technology makes use of the easy and efficient collection of beads in magnetic field to facilitate antibody purification in a simple workflow of “bind-wash-elute”. In contrast to common separation techniques, this method does not require columns or centrifugation, and is therefore ideal in high-throughput applications. ;Immobilized 40μg S protein trimer/1mg beads can bind the Anti-S1 Antibody with an EC50 of 0.5720μg/mL (QC tested).