Learn More
ACROBiosystems ACROBiosystems SARS-CoV-2 Spike Trimer (T95I, G142D, E154K, L452R, E484Q, D614G, P681R, Q1071H), His Tag (MALS verified)
Supplier:  ACROBiosystems SPNC52HR50UG
Encompass_Preferred
Recombinant Protein;50UG;SARS-CoV-2 Spike Trimer, His Tag (SPN-C52Hr) is expressed from human 293 cells (HEK293). It contains AA Val 16 - Pro 1213 (Accession # QHD43416.1). The mutations (T95I, G142D, E154K, L452R, E484Q, D614G, P681R) were identified in the SARS-CoV-2 variants which emerged in India (known as B.1.617). Proline substitutions (F817P, A892P, A899P, A942P, K986P, V987P) and alanine substitutions (R683A and R685A) are introduced to stabilize the trimeric prefusion state of SARS-CoV-2 S protein and abolish the furin cleavage site, respectively.;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 138.2 kDa. The protein migrates as 150-200 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized SARS-CoV-2 Spike Trimer, His Tag (Cat. No. SPN-C52Hr) at 1 μg/mL (100 μL/well) can bind Human ACE2, Fc Tag (Cat. No. AC2-H5257) with a linear range of 0.8-25 ng/mL (QC tested).;Immobilized SARS-CoV-2 Spike Trimer, His Tag
Catalog No. 50-210-7018