Learn More
ACROBiosystems ACROBiosystems SARS-CoV-2 Spike Trimer (G75V, T76I, SYLTPGD 247-253 del, L452Q, F490S, D614G, T859N), His Tag
Supplier:  ACROBiosystems SPNC52HS500UG
Encompass_Preferred
Recombinant Protein;500UG;SARS-CoV-2 Spike Trimer, His Tag (SPN-C52Hs) is expressed from human 293 cells (HEK293). It contains AA Val 16 - Pro 1213 (Accession # QHD43416.1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 137.2 kDa.
Catalog No. 50-210-6819